Learn More
Invitrogen™ Goat anti-Mouse IgG1 Secondary Antibody, HRP
Descripción
This antibody does not react with human. Goat anti Mouse IgG1 antibody recognizes Mouse IgG1.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones
Especificaciones
| Antígeno | Mouse IgG1 |
| Aplicaciones | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | HRP |
| Formulación | PBS with 50% glycerol and 0.0095% Methylisothiazolone; pH 7.4 |
| Especie del huésped | Goat |
| Inmunógeno | Mouse IgG1 paraproteins. |
| Método de purificación | Affinity chromatography |
| Cantidad | 1 mL |
| Estado normativo | RUO |
| Mostrar más |