Learn More
Invitrogen™ Goat anti-Human IgG, IgM, IgA (H+L) Secondary Antibody
Código de producto. 11855875
Change view
Click to view available options
Cantidad:
2 mg
Tamaño de la unidad:
2 miligramos
11855875 2 mg 2 miligramos
1 options
Código de producto. 11855875 Proveedor Invitrogen™ N.º de proveedor 31128
Goat Polyclonal Secondary Antibody
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31128 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31128 reacts with the heavy chains of human IgA, IgG, and IgM, and with the light chains common to most human immunoglobulins. The antibody does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Human IgG, IgM, IgA (H+L)
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 2.4 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative; pH 7.6
Especie del huésped Goat
Método de purificación Antigen affinity chromatography
Cantidad 2 mg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG