Learn More
Goat anti-Human IgA Heavy Chain Secondary Antibody, DyLight 650, Novus Biologicals™
Goat Polyclonal Antibody
Marca:  Novus Biologicals NBP1-75754
Código de producto. 18530111
  • ONLINE EXCLUSIVE PRICE
    241.00€
    227.85€ / 1 miligramo
    Ahorro 13.15€
    5% Descuento
Descripción
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
IgA Heavy Chain
Polyclonal
DyLight 650
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Purified human IgA, (Alpha chain)
1 mg
Adaptive Immunity, Immunology
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
IgG
Flow Cytometry, Immunomicroscopy
LYOPH
Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Goat
Affinity purified
RUO
Secondary
Human
Purified
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones