Learn More
CXCL14, Goat, Polyclonal Antibody, Abnova™
Descripción
This gene belongs to the cytokine gene family which encode secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC (Cys-X-Cys) subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes but not for lymphocytes, dendritic cells, neutrophils or macrophages. It has been implicated that this cytokine is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | CXCL14 |
| Aplicaciones | ELISA |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Descripción | Goat polyclonal antibody raised against synthetic peptide of CXCL14. |
| Dilución | ELISA (1:32000) The optimal working dilution should be determined by the end user. |
| Formulación | In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) |
| génica | CXCL14 |
| N.º de referencia del gen | NP_004878.2 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.