Learn More
AFP Goat anti-Human, Polyclonal Antibody, Abnova™
Descripción
This gene encodes alpha-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | AFP |
| Aplicaciones | ELISA |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Goat polyclonal antibody raised against native AFP. |
| Dilución | The optimal working dilution should be determined by the end user. |
| Formulación | In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide) |
| génica | AFP |
| Alias de gen | FETA/HPAFP |
| Símbolos de los genes | AFP |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.