Learn More
ABCC4 Goat anti-Human, Rat, Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | ABCC4 |
| Aplicaciones | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Descripción | Goat polyclonal antibody raised against synthetic peptide of ABCC4. |
| Dilución | ELISA (1:32000) Western Blot (1-3 ug/mL) Immunohistochemistry (2-4 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulación | In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) |
| génica | ABCC4 |
| N.º de referencia del gen | NP_005836.2 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.