missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13-372.14), Invitrogen™

Product Code. p-7244903
Change view
Click to view available options
Cantidad:
1 mg
100 μg
500 μg
Unit Size:
1 miligramo
100 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
17820496 100 μg 100 microgramos
11573423 500 μg 1 miligramo
17890176 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17820496 Supplier Invitrogen™ Supplier No. MA546088

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10815 detects Gonadotropin Releasing Hormone from human samples. MA1-10815 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10815 immunogen is purified human gonadotropin releasing hormone.

The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GnRH1
Aplicaciones ELISA, Immunohistochemistry
Clasificación Monoclonal
Clon LHRH13-372.14
Concentración 1.40 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative; pH 7.4
génica GNRH1
N.º de referencia del gen P01148
Alias de gen Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
Símbolos de los genes GNRH1
Especie del huésped Mouse
Inmunógeno Purified human gonadotropin releasing hormone.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2796
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.