Learn More
GMEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16881454 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16881454 20 μL 20 microlitros
16871454 150 μL 150 microlitros
2 options
Código de producto. 16881454
Marca: Proteintech 133141AP20UL
Rabbit Polyclonal Antibody
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB40, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Single nucleotide polymorphisms near ZBTB40 have been linked to differences in bone mineral density of the hip and lumbar spine, and had a significant association with low-trauma fractures suggesting that ZBTB40 may be involved in bone development and formation.
Especificaciones
Antígeno GMEB2
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica GMEB2
N.º de referencia del gen O88873, P58929, Q9UKD1
Alias de gen DNA binding protein p79PIF, GMEB 2, GMEB2, KIAA1269, P79PIF, PIF p79, PIF79
Símbolos de los genes Gmeb2
Especie del huésped Rabbit
Inmunógeno GMEB2 Fusion Protein Ag4157
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 229004, 26205, 83635
Especies diana Mouse, Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG