missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine Synthetase Antibody (GLUL/6600) [Janelia Fluor™ 585], Novus Biologicals™
Descripción
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Bovine,Equine,Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Glutamine Synthetase |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | GLUL/6600 |
| Conjugado | Janelia Fluor 585 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant fragment (around aa 50-250) of human Glutamine Synthetase protein (exact sequence is proprietary) (Uniprot: P15104) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?