Learn More
Glutamine Synthetase Antibody (GLUL/6600) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP3-14023JF525
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Bovine,Equine,Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
| Glutamine Synthetase | |
| Monoclonal | |
| Janelia Fluor 525 | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| Recombinant fragment (around aa 50-250) of human Glutamine Synthetase protein (exact sequence is proprietary) (Uniprot: P15104) | |
| 0.1 mL | |
| Hypoxia, Neurodegeneration, Neuroscience, Vision | |
| 2752 | |
| Store at 4°C in the dark. | |
| IgG2bc κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| GLUL/6600 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Equine, Human | |
| Purified |