Learn More
GLUT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 20403-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16869934
  • 410.00€ / 150 microlitros
Descripción
This antibody was generated against the internal region of human GLUT3 and may cross-react with GLUT14.20403-1-AP antibody detects the bands around 48-60 kDa in SDS-PAGE.
Glucose is the major source of our energy and there are numerous isoforms of the glucose transporter in mammals, including Glut1, Glut2, Glut3, Glut4, Glut5, Glut6, Glut7, Glut8 and Glut9. The Glut5 gene located on the short arm of human chromosome 1 encodes a 501-amino acid facilitative glucose transporter. Glut5 mRNA is highly expressed in small intestine and to a lesser extent in kidney, skeletal muscle and adipose tissue. Glut5 plays a critical role in fructose absorption in the small intestine and its expression is highly induced when exposed to a fructose-enriched diet. Glut5 transporter expressed in human skeletal muscle is specifically localized to the plasma membrane, where it participates in regulating hexose transfer across the sarcolemma. Glut8, a novel glucose transporter-like protein, exhibits significant sequence similarity with the other members of sugar transporter family. Glut8 comprises 12 putative membrane-spanning helices and several conserved motifs, which are important for transport activity. In human tissues, Glut8 is predomitly expressed in testis and, to a lesser extent, in most other tissues including skeletal muscle, heart, small intestine and brain.
Especificaciones
Especificaciones
GLUT3
Polyclonal
Unconjugated
SLC2A3
GLUT 3, GLUT3, SLC2A3
Rabbit
Antigen Affinity Chromatography
RUO
20527, 6515
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11169, P32037
SLC2A3
GLUT3 Fusion Protein Ag14203
150 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones