Learn More
GLUT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16879934 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16879934 20 μL 20 microlitros
16869934 150 μL 150 microlitros
2 options
Código de producto. 16879934
Marca: Proteintech 204031AP20UL
Rabbit Polyclonal Antibody
This antibody was generated against the internal region of human GLUT3 and may cross-react with GLUT14.20403-1-AP antibody detects the bands around 48-60 kDa in SDS-PAGE.
Glucose is the major source of our energy and there are numerous isoforms of the glucose transporter in mammals, including Glut1, Glut2, Glut3, Glut4, Glut5, Glut6, Glut7, Glut8 and Glut9. The Glut5 gene located on the short arm of human chromosome 1 encodes a 501-amino acid facilitative glucose transporter. Glut5 mRNA is highly expressed in small intestine and to a lesser extent in kidney, skeletal muscle and adipose tissue. Glut5 plays a critical role in fructose absorption in the small intestine and its expression is highly induced when exposed to a fructose-enriched diet. Glut5 transporter expressed in human skeletal muscle is specifically localized to the plasma membrane, where it participates in regulating hexose transfer across the sarcolemma. Glut8, a novel glucose transporter-like protein, exhibits significant sequence similarity with the other members of sugar transporter family. Glut8 comprises 12 putative membrane-spanning helices and several conserved motifs, which are important for transport activity. In human tissues, Glut8 is predomitly expressed in testis and, to a lesser extent, in most other tissues including skeletal muscle, heart, small intestine and brain.
Especificaciones
Antígeno GLUT3
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.33 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica SLC2A3
N.º de referencia del gen P11169, P32037
Alias de gen GLUT 3, GLUT3, SLC2A3
Símbolos de los genes SLC2A3
Especie del huésped Rabbit
Inmunógeno GLUT3 Fusion Protein Ag14203
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20527, 6515
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG