missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucuronic Acid Epimerase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP2-31985
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Glucuronic Acid Epimerase Polyclonal specifically detects Glucuronic Acid Epimerase in Human, Equine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| Glucuronic Acid Epimerase | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| O94923 | |
| GLCE | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SDKAIQFPRRSSSGFRVDGFEKRAAASESNNYMNHVAKQQSEEAFPQEQQKAPPVVGGFNSNVGSKVLGLKYEEIDCLINDEHTIKGRREGNEVFLPFTWVEKYFDVYGKVVQYDGYDR | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| D-glucuronyl C5-epimerase, EC 5.1.3, EC 5.1.3.-, glucuronic acid epimerase, glucuronyl C5-epimerase, Heparin/heparan sulfate:glucuronic acid C5-epimerase, HSEPIheparin/heparan sulfate-glucuronic acid C5-epimerase, KIAA0836heparan sulfate epimerase, UDP-glucuronic acid epimerase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26035 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido