Learn More
Laboratories Sales™ Glass Snap Top Vials
Marca: Laboratories Sales™ 105000
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Manufactured from type 1 neutral glass
- Particularly suitable for the storage of powders and dry samples
- Snap ring neck finish fits a flip off style, flexible low density polythene closure supplied separately
- Single cap fits all vial sizes