Learn More
Marienfeld Superior™ Glass Capillary Tubes
Use Marienfeld Superior™ Glass Capillary Tubes to determine melting points during laboratory experimentation.
Marca:  Marienfeld Superior™ 2930208
Código de producto. 10018677
  • 76.13€ / paquete de 1000
Descripción
  • Double-sided
  • 80mm * 0.6mm
  • Pack = 1000 pieces