missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GJB6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24215-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Gap junctions allow the transport of ions and metabolites between the cytoplasm of adjacent cells. They are formed by two hemichannels, made up of six connexin proteins assembled in groups. Each connexin protein has four transmembrane segments, two extracellular loops, a cytoplasmic loop formed between the two inner transmembrane segments, and the N- and C-terminus both being in the cytoplasm. The specificity of the gap junction is determined by which connexin proteins comprise the hemichannel. In the past, connexin protein names were based on their molecular weight, however the new nomenclature uses sequential numbers based on which form of the gap junction is present. This gene encodes one of the connexin proteins. Mutations in this gene have been found in some forms of deafness and in some families with hidrotic ectodermal dysplasia.Especificaciones
| GJB6 | |
| Polyclonal | |
| Unconjugated | |
| GJB6 | |
| Connexin 30, CX30, DFNA3, DFNA3B, DFNB1B, ED2, EDH, Gap junction beta 6 protein, GJB6, HED | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10804 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95452 | |
| GJB6 | |
| GJB6 Fusion Protein Ag21529 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido