missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GINS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA584331
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: ELVTIIPNFS LDKIYLIGGD LGPFNPGLPV EVPLWLAINL KQRQKCRLLP PEWMDVEKLE KMRDHERKEE TFTPMPSPYY MELTKLLLNH ASDNIPKADE IRTLVKDMWD TRIAKLRVSA DSFVRQQEAH AKLDNLTLME INTSG Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 92%.
The yeast heterotetrameric GINS complex is made up of Sld5, Psf1, Psf2, and Psf3. The formation of this complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts (Ueno et al., 2005 ). See GINS1 for additional information about the GINS complex.
Especificaciones
| GINS2 | |
| Polyclonal | |
| Unconjugated | |
| GINS2 | |
| 2210013I18Rik; 4833427B12Rik; AI323585; CGI-122; DC5; DNA replication complex GINS protein PSF2; GINS complex subunit 2; GINS complex subunit 2 (Psf2 homolog); Gins2; HSPC037; Pfs2; PSF2; RGD1311055 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51659 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y248 | |
| GINS2 | |
| Recombinant protein corresponding to Human GINS2. Recombinant protein control fragment (Product #RP-88827). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido