missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GINS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA562341
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: HMAAEEMEWF NNYKRSLATY MRSLGGDEGL DITQDMKPPK SLYIEVRCLK DYGEFEVDDG TSVLLKKNSQ HFLPRWKCEQ LIRQGVLEH Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
The yeast heterotetrameric GINS complex is made up of Sld5 (GINS4; MIM 610611), Psf1, Psf2 (GINS2; MIM 610609), and Psf3 (GINS3; MIM 610610). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
Especificaciones
| GINS1 | |
| Polyclonal | |
| Unconjugated | |
| GINS1 | |
| 2810418N01Rik; DNA replication complex GINS protein PSF1; GINS complex subunit 1; GINS complex subunit 1 (Psf1 homolog); GINS complex subunit 4 (Sld5 homolog); Gins1; Gins4; KIAA0186; mKIAA0186; partner of sld five-1; PSF1; RP4-691N24.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9837 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14691 | |
| GINS1 | |
| Recombinant protein corresponding to Human GINS1. Recombinant protein control fragment (Product #RP-99313). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido