missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGCX Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA519155
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
GGCX (gamma-glutamyl carboxylase), also known as GC or VKCFD1 (Vitamin K-dependent gamma-carboxylase), is a 758 amino acid multi-pass membrane protein. Localized to the membrane of the endoplasmic reticulum, GGCX functions to mediate the vitamin K-dependent carboxylation of glutamate residues on target proteins, thereby producing calcium binding gamma-carboxyglutamate (Gla) residues on these proteins and simultaneously converting vitamin K to vitamin K epoxide. GGCX exists as a monomer and, via its ability to modify glutamate residues, it accomplishes the post-translational changes that are necessary for the activity of all vitamin K-dependent proteins (such as blood coagulation and bone matrix proteins). Defects in the gene encoding GGCX are the cause of combined deficiency of vitamin K-dependent clotting factors 1 (VKCFD1) and PXE-like disorder with multiple coagulation factor deficiency, both of which are characterized by abnormal skin, blood or bone function.
Especificaciones
| GGCX | |
| Polyclonal | |
| Unconjugated | |
| Ggcx | |
| FLJ26629; Gamma-glutamyl carboxylase; GC; GGCX; Peptidyl-glutamate 4-carboxylase; vitamin K gamma glutamyl carboxylase; Vitamin K-dependent gamma-carboxylase; VKCFD1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2677 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P38435 | |
| Ggcx | |
| Synthetic peptide sequence (PPESNPDPVHSE) corresponding to the C-terminus amino acids of GGCX (aa 746-758). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido