Learn More
Invitrogen™ GFR alpha-2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA118399
Código de producto. 11532042
  • 578.00€ / 100 microlitros
Descripción
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
GFR alpha-2
Polyclonal
Unconjugated
GFRA2
GDNF family receptor alpha 2; GDNF family receptor alpha-2; GDNF re; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; Gdnfrb; GDNFR-beta; Gfra2; GFR-alpha 2; GFR-alpha-2; glial cell line derived neurotrophic factor family receptor alpha 2; glial cell line derived neurotrophic factor receptor, beta; HpV18gp1; neurturin receptor alpha; NRTNR-alpha; NTNRA; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; RETL2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored; TRNR2; tyrosine kinase receptor ligand 2
Rabbit
100 μL
Primary
Human, Rat
Antibody
IgG
Immunohistochemistry (Frozen), Western Blot
Conc. Not Determined
whole serum with no preservative
O00451
GFRA2
Synthetic peptide corresponding to residues P(376) R V E K T P S L P D D L S D(390) of human GFR alpha-2.
RUO
25136, 2675
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones