missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFP Polyclonal Antibody, Alexa Fluor™ 555
GREENER_CHOICE

Código de producto. 10719194
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10719194 100 μL cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10719194 Proveedor Invitrogen™ N.º de proveedor A31851

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

At the time of preparation, the products are certified to be free of unconjugated dyes and are tested in a cytological experiment to ensure low nonspecific staining. Several Alexa Fluor™ dye-conjugates made from the rabbit anti-GFP IgG fraction are also available. The Alexa Fluor™ dyes provide for extraordinarily bright antibody conjugates. The approximate fluorescence excitation (Ex) and emission maxima for this Alexa Fluor™ 555 conjugate are 555 nm and 565 nm, respectively. These anti-GFP antibody conjugates are suited for detection of native GFP, GFP variants, and most GFP-fusion proteins by western blot analysis and immunocytochemistry. For initial experiments, we recommend trying dilutions ranging from 1:200 to 1:2,000 with our fluorophore-labeled antibodies, a final concentration of 1-10 μg/mL should be satisfactory for most immunocytochemical applications. If using the rabbit anti-GFP antibody-Alexa Fluor™ conjugates in western blotting, dilute the conjugate 1:2,000 in PBS to obtain a final antibody concentration of 1 μg/mL. HeLa and U2-OS cells were tested with this antibody in immunocytochemistry, but not with paraffin-embedded sections. It is a good practice to centrifuge the protein conjugate solutions briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step eliminates any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.

The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno GFP
Aplicaciones ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 2 mg/mL
Conjugado Alexa Fluor 555
Formulación PBS with 5mM sodium azide; pH 7.5
Alias de gen GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP
Especie del huésped Rabbit
Inmunógeno The GFP was isolated directly from the jellyfish Aequorea victoria.
Método de purificación Ion-exchange chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
Especies diana Tag
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Molécula diana Tag
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.