missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFP Monoclonal Antibody (GF28R), DyLight™ 650
GREENER_CHOICE

Código de producto. 15216189
Change view
Click to view available options
Cantidad:
50 μL
Tamaño de la unidad:
50 microgramos
Código de producto. Cantidad unitSize
15216189 50 μL 50 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15216189

Marca: Invitrogen™ MA515256D650

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-15256-D650 has been successfully used for immunofluorescence. DyLight 650 has an excitation/emission of 652/672 nm.

The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno GFP
Aplicaciones Immunocytochemistry
Clasificación Monoclonal
Clon GF28R
Concentración 1 mg/mL
Conjugado DyLight 650
Formulación PBS with proprietary stabilizer and 0.02% sodium azide
Alias de gen GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP
Especie del huésped Mouse
Inmunógeno GFP from the jellyfish Aequorea Victoria N-terminal peptide-KLH conjugated.
Método de purificación Protein A
Cantidad 50 μL
Estado normativo RUO
Primario o secundario Primary
Especies diana Tag
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Molécula diana Tag
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.