missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFI1 Polyclonal Antibody

Descrizione
GFI1 Polyclonal Antibody for Western Blot, IHC (P)
GFI1 is a nuclear zinc finger (ZF) transcriptional repressor that plays an important role in hematopoiesis and inner ear development, and has been implicated in lymphomagenesis. The Gfi1 protein expression is regulated by the ubiquitin-proteasome system. It is reported that ubiquitin ligase Triad1 interacts with the DNA-binding domain of Gfi-1. Gfi-1 represses transcription by directly binding to the consensus DNA sequence in the promoters of its target genes. Deficiency of Gfi-1 causes neutropenia and an accumulation of granulomonocytic precursor cells that is reminiscent of a myelodysplastic syndrome in mice.
Specifica
Specifica
| Antígeno | GFI1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% sodium azide |
| génica | GFI1 |
| N.º de referencia del gen | P70338, Q07120, Q99684 |
| Alias de gen | AW495828; FLJ94509; Gfi1; GFI-1; GFI1A; growth factor independence-1; growth factor independent 1; growth factor independent 1 transcription repressor; growth factor independent 1 transcriptional repressor; growth factor independent protein 1; Growth factor independent-1; Pal1; Pal-1; SCN2; Zinc finger protein 163; zinc finger protein Gfi-1; ZNF163 |
| Símbolos de los genes | GFI1 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?