missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP (Glial Fibrillary Acidic Protein) Mouse, anti-Human, Unconjugated, Clone: 2E1.E9, STEMCELL Technologies
Descripción
- The 2E1.E9 antibody reacts with glial fibrillary acidic protein (GFAP), an ∼49 kDa type III intermediate filament (IF) protein that, within the central nervous system, is expressed primarily by astrocytes, though found at high levels in some glial-derived tumors
- GFAP is thought to contribute to the structural architecture and strength of the cytoskeleton
- The 2E1.E9 antibody does not cross-react with other IF proteins and can be used to distinguish astrocytes from other glial cells
- GFAP has also been identified in Leydig cells, keratinocytes, chondrocytes and osteocytes
- Many mutations in the GFAP gene (>50) have been associated with Alexander disease, a progressive leukoencephalopathy characterized by cytoplasmic inclusions and dysfunctional myelination
- This antibody clone has been verified for labeling neural stem and progenitor cells grown in NeuroCult™ NS-A Proliferation Kit (Human; Catalog #05751) and NeuroCult™ Proliferation Kit (Mouse; Catalog #05702)
Especificaciones
Especificaciones
| Antígeno | GFAP (Glial Fibrillary Acidic Protein) |
| Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 2E1.E9 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 100 μg |
| Primario o secundario | Primary |
| Especies diana | Human, Rat, Mouse |
| Isotype | IgG2b |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?