Learn More
GFAP (Astrocyte & Neural Stem Cell Marker) Mouse, Clone: GFAP/2076, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 2670-MSM6-P1
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| GFAP (Astrocyte & Neural Stem Cell Marker) | |
| Cytoplasmic | |
| GFAP/2076 | |
| Unconjugated | |
| Mouse | |
| ∽50kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 2670 | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human GFAP protein fragment (apprx. aa 101-200) | |
| Brain or Astrocytoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |