missing translation for 'onlineSavingsMsg'
Learn More

GFAP Antibody (GFAP/8616R), CoraFluor™ 1, Novus Biologicals™

Código de producto. 30141271
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
30141271 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30141271

Brand: Novus Biologicals (Bio-Techne) NBP324136CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GFAP
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon GFAP/8616R
Conjugado CoraFluor 1
Formulación PBS
Alias de gen ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Especie del huésped Rabbit
Inmunógeno Recombinant fragment (around aa150-350) of human GFAP protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 2670
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.