missing translation for 'onlineSavingsMsg'
Learn More

Genipin 1-β-D-gentiobioside, MedChemExpress

Código de producto. 16459258
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Unit Size:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16459258 10 mg 10 miligramos
16421678 5 mg 5 miligramos
16413946 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16459258 Supplier Medchem Express Supplier No. HYN2094

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Genipin 1-β-D-gentiobioside (Genipin 1-gentiobioside) is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis, which possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.

Genipin 1-β-D-gentiobioside (Genipin 1-gentiobioside) is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis, which possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.

Specifications

Nombre del producto químico o material Genipin 1-β-D-gentiobioside
CAS 29307-60-6
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C23H34O15
Cantidad 10 mg
Sinónimo Genipin 1-gentiobioside Genipin 1-β-gentiobioside Genipin gentiobioside
Información de solubilidad Ethanol : 125 mg/mL (227.06 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (181.65 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO)([H])O[C@H]3[C@@](C(CO)=CC4)([H])[C@@]4([H])C(C(OC)=O)=CO3
Peso molecular (g/mol) 550.51
Formula Weight (peso de la fórmula) 550.51
Porcentaje de pureza 98.65%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.