Learn More
Thermo Scientific™ Kit de ensayo GeneBLAzer™ EDNRB HEK 293T DA
Código de producto. 10484035
Change view
Click to view available options
Cantidad:
1 placa
Tamaño de la unidad:
cada uno
10484035 1 placa cada uno
1 options
Código de producto. 10484035
Marca: Thermo Scientific™ K1349
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
GeneBLAzer™ EDNRB HEK 293T DA (Division Arrested) cells and EDNRB-NFAT-bla HEK 293T cells contain the human Endothelin type B (EDNRB) receptor stably integrated into the CellSensor™ NFAT-bla HEK 293T cell line.
This cell line is available as dividing cells by requesting a quote for K1224 from dividingcells@invitrogen.com
GeneBLAzer™ EDNRB HEK 293T DA (Division Arrested) cells and EDNRB-NFAT-bla HEK 293T cells contain the human Endothelin type B (EDNRB) receptor stably integrated into the CellSensor™ NFAT-bla HEK 293T cell line. CellSensor™ NFAT-bla HEK 293T cells (Cat. No. K1129) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells(NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.
DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both EDNRB HEK 293T DA cells and EDNRB-NFAT-bla HEK 293T cells are functionally validated for Z'-factor and EC50 concentrations of ET-1 (Figure 1). In addition, EDNRB-NFAT-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
Endothelin Receptor Type B (ENDRB) mediates signaling of endothelin proteins that regulate several critical biological processes, including the development and function of blood vessels, the production of certain hormones, and the stimulation of cell growth and division. Defects in EDNRB mediated signaling have been implicated in a wide range of diseases, including cancer, hypertension and cardiovascular disorders.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso NM_000115.1
Entrada del ensayo Gen informador de beta lactamasa a base de células
Estado de célula Células de división detenida
Contenido y almacenamiento Nitrógeno líquido
Descripción Kit de ensayo GeneBLAzer™ EDNRB HEK 293T DA, 1 placa, cantidad de células ∽ 2 400 000 células/vial, línea de células 293T (HEK), método de detección fluorescente, objetivo quimiomodulable GPCR, indicador de BLA (beta-lactamasa), almacenamiento en nitrógeno líquido
Método de detección Fluorescente
Para utilizar con (aplicación) Adquisición de imágenes celulares, farmacología y biofarmacología, análisis celular, adquisición de imágenes celulares, ensayos celulares con indicador, ensayos celulares y de beta-lactamasa, descubrimiento y desarrollo de fármacos, identificación y validación de objetivo y potencial, biología de receptores acoplados a proteínas G, ensayos celulares con indicadores GPCR, ensayos ADME/Tox basados en dianas, ensayos de adme celular, ensayos de adme del receptor acoplado de la proteína G
ID de gen (Entrez) 1910
Símbolo de gen EDNRB
Incluye Kit de ensayo GeneBLAzer™ EDNRB HEK 293T DA. Cada sistema contiene suficientes células de división detenida (DA) y sustrato para ensayar 1 placa de 384 pocillos. (Otros materiales se requieren por separado; consulte el protocolo). Incluye: Células EDNRB HEK 293T DA,™kit de carga LiveBLazer -FRET B/G, 70 μg, solución D, 1 ml, 2 protocolos, certificado de análisis
Estado de la población Origen del clon unicelular
Línea de productos GeneBLAzer
Cantidad 1 placa
Gen marcador BLA (betalactamasa)
Tipo de muestra 293T (HEK)
Entrada diana EDNRB
Tipo Kit de ensayo GeneBLAzer™ EDNRB HEK 293T DA
Línea de células 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Mostrar más Mostrar menos
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.