missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gelsolin Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA529910
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: H1299, HeLa, human plasma, mouse plasma, rat plasma. Predicted reactivity: Mouse (93%), Rat (90%), Chicken (80%), Sheep (96%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene binds to the 'plus' ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
Especificaciones
| Gelsolin | |
| Polyclonal | |
| Unconjugated | |
| GSN | |
| Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 227753, 2934, 296654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with no preservative | |
| P06396, P13020, Q68FP1 | |
| GSN | |
| Recombinant protein encompassing a sequence within the center region of human Gelsolin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido