missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDF9 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15645025
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15645025 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15645025 Supplier Invitrogen™ Supplier No. PA551102

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GDF9 Polyclonal Antibody detects endogenous levels of GDF9 protein.

GDF9 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues.Growth factors synthesized by ovarian somatic cells directly affect oocyte growth and function. GDF9 is expressed in oocytes and is thought to be required for ovarian folliculogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GDF9
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
génica GDF9
N.º de referencia del gen O60383, Q07105
Alias de gen Gdf9; Gdf-9; growth differentiation factor 9; growth/differentiation factor 9
Símbolos de los genes GDF9
Especie del huésped Rabbit
Inmunógeno Synthesized peptide derived from the C-terminal region of human GDF-9.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 14566, 2661, 59304
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.