Learn More
GDF15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. p-7212419 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16825635 20 μL 20 microlitros
16815635 150 μL 150 microlitros
2 options
Código de producto. 16825635
Marca: Proteintech 274551AP20UL
Rabbit Polyclonal Antibody
Bone morphogenetic proteins (e. g., BMP5, MIM 112265) are members of the transforming growth factor-beta (see TGFB1, MIM 190180) superfamily and regulate tissue differentiation and maintece. They are synthesized as precursor molecules that are processed at a dibasic cleavage site to release C-terminal domains containing a characteristic motif of 7 conserved cysteines in the mature protein.
Especificaciones
Antígeno GDF15
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica GDF15
N.º de referencia del gen Q99988, Q9Z0J7
Alias de gen GDF 15, GDF15, MIC 1, MIC1, NAG 1, NRG 1, NSAID activated gene 1 protein, NSAID regulated gene 1 protein, PDF, PLAB, Placental TGF beta, PTGFB
Símbolos de los genes GDF15
Especie del huésped Rabbit
Inmunógeno GDF15 Fusion Protein Ag26760
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23886, 9518
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG