missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Antibody detects endogenous levels of total TUBGCP3.
The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP3 (gamma-tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is an ubiquitously expressed 907 amino acid member of the gamma-Tubulin complex.
Especificaciones
Especificaciones
| Antígeno | GCP3 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | TUBGCP3 |
| N.º de referencia del gen | P58854, Q96CW5 |
| Alias de gen | 104p; gamma-ring complex protein 104 kDa; Gamma-tubulin complex component 3; GCP3; GCP-3; Grip104; h104p; hGCP3; hGrip104; hSpc98; SPBC98; Spc98; Spc98p; spindle pole body protein; spindle pole body protein Spc98 homolog; Tubgcp3; tubulin gamma complex associated protein 3; tubulin, gamma complex associated protein 3 |
| Símbolos de los genes | TUBGCP3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
