Descripción
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
Especificaciones
Specifications
| Antígeno | GBX2 |
| Aplicaciones | Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.33 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | GBX2 |
| N.º de referencia del gen | P48031, P52951 |
| Alias de gen | gastrulation brain homeobox 2, GBX2, Homeobox protein GBX 2 |
| Símbolos de los genes | Gbx2 |
| Show More |