missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GATM Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16382345
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
16382345 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16382345

Marca: Invitrogen™ PA585409

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (97%), Zebrafish (87%), Xenopus laevis (91%), Dog (95%), Pig (94%), Chicken (91%), Bovine (94%). Positive Control: HeLa, HepG2, mouse liver, human GATM-transfected HEK293 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

AGAT, also known as glycine amidinotransferase (L-arginine:glycine amidinotransferase), GATM or transamidinase, is a 423 amino acid protein belonging to the amidinotransferase family. Encoded by a gene that maps to human chromosome 15q21.1, AGAT exists as three alternatively spliced isoforms and consists of a homodimer, with equilibrium between monomeric and dimeric forms favoring a monomer subunit structure. AGAT localizes to mitochondrial inner membranes, peripheral membranes and cytoplasm. Biallelically expressed in placenta and fetal tissues, AGAT is also expressed in brain, heart, liver, lung, salivary gland and skeletal muscle tissue, with high expression in kidney. AGAT is elevated in the myocardium during heart failure and is decreased in inter-uterine growth restriction (IUGR)-associated placenta. AGAT catalyzes biosynthesis of guanidinoacetate, the immediate precursor of creatine, which plays a vital role in energy metabolism in muscle tissues. AGAT defects are associated with arginine:glycine amidinotransferase deficiency, an autosomal recessive disorder characterized by developmental delay or regression, mental retardation, severe disturbance of expressive and cognitive speech and severe depletion of creatine/phosphocreatine in brain. AGAT may be linked to embryonic and central nervous system development and may function in heart failure response by elevating local creatine synthesis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno GATM
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
génica GATM
N.º de referencia del gen P50440, Q9D964
Alias de gen 1810003P21Rik; Agat; AI314789; AT; CCDS3; GATM; glycine amidinotransferase; glycine amidinotransferase (L-arginine:glycine amidinotransferase); glycine amidinotransferase, mitochondrial; L-arginine: glycine amidinotransferase; l-arginine:glycine amidinotransferase; testicular secretory protein Li 19; transamidinase
Símbolos de los genes GATM
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human GATM. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2628, 67092
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.