Learn More
BOLA Gas Inlet Tubes
Descripción
- For constructing a gas washing equipment.
- Tube with inner diameter 5 mm, one side with hose connector dia. 9 mm for connecting tubing, other side with thread M 8x1 for connecting a gas frit or a gas distributor.
Especificaciones
Especificaciones
| Longitud (métrico) | 600 mm |
| Esterilizables en autoclave | Autoclavable |