missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAD65/GAD67 Polyclonal Antibody

Descripción
This antibody detects endogenous protein at a molecular weight of 65 and 67 kDa. Purity is >95% by SDS-PAGE.
GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA ( gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD-65 will transiently activate to assist in GABA production. The loss of GAD-65 is detrimental and can impair GABA neurotransmission, however the loss of GAD-67 is lethal. Due to alternative splicing, two isoforms exist for GAD-67: the predominant GAD-67 form and the minor GAD-25 form.
Especificaciones
Especificaciones
| Antígeno | GAD65/GAD67 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | GAD2 |
| N.º de referencia del gen | P18088, P48318, P48320, Q05329, Q05683, Q99259 |
| Alias de gen | 6330404F12Rik; 65 kDa glutamic acid decarboxylase; 67 kDa glutamic acid decarboxylase; CPSQ1; EC 4.1.1.15; EP10; FLJ45882; GAD; GAD 1; GAD 2; GAD 65; GAD 67; GAD(65); GAD1; Gad-1; Gad2; Gad-2; GAD25; GAD44; Gad65; GAD-65; GAD67; GAD-67; Glutamate decarboxylase 1; Glutamate decarboxylase 1 (brain); glutamate decarboxylase 1 (brain, 67kDa); glutamate decarboxylase 1 variant GAD67NT; glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase 67 kDa isoform; GLUTAMATE DECARBOXYLASE, 67 KD ISOFORM (GAD-67) (67 KD GLUTAMIC ACID DECARBOXYLASE); Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase; glutamic acid decarboxylase 1; glutamic acid decarboxylase 2; glutamic acid decarboxylase 65; SCP |
| Símbolos de los genes | Gad1, GAD2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?