missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GABRE Polyclonal Antibody

Código de producto. 15514572
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15514572 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15514572

Marca: Invitrogen™ PA541023

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Peptide sequence: DVVYGPQPQP LENQLLSEET KSTETETGSR VGKLPEASRI LNTILSNYDH Sequence homology: Guinea Pig: 85%; Human: 100%; Mouse: 86%; Rabbit: 93%.

The product of this gene belongs to the ligand-gated ionic channel (TC 1.A.9) family. It encodes the gamma-aminobutyric acid (GABA) A receptor which is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes an epsilon subunit. It is mapped to chromosome Xq28 in a cluster comprised of genes encoding alpha 3, beta 4 and theta subunits of the same receptor. Alternatively spliced transcript variants have been identified, but only one is thought to encode a protein.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GABRE
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica Gabre
N.º de referencia del gen P78334
Alias de gen GABA(A) mu Receptor; GABA(A) receptor; GABA(A) receptor subunit epsilon; GABA(A) receptor, epsilon; GABRE; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, subunit epsilon; gamma-aminobutyric acid (GABA-A) receptor, subunit epsilon; gamma-aminobutyric acid A receptor, epsilon; gamma-aminobutyric acid receptor subunit epsilon; gamma-aminobutyric acid type A receptor epsilon subunit; gamma-aminobutyric acid type A receptor subunit epsilon
Símbolos de los genes Gabre
Especie del huésped Rabbit
Inmunógeno Synthetic peptide directed towards the N-terminal of human GABRE (aa 35-84).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2564
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.