Learn More
G6PD Mouse anti-Human, Unconjugated, Clone: 2A7B12, Proteintech
Código de producto. 16818195 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16818195 20 μL 20 microlitros
16808195 150 μL 150 microlitros
2 options
Código de producto. 16818195
Marca: Proteintech 663731IG20UL
Mouse Monoclonal Antibody
The G6PD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human G6PD. This antibody recognizes human, mouse, rat antigen. The G6PD antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno G6PD
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Monoclonal
Clon 2A7B12
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 2539
Especies diana Human, Rat, Mouse
Isotype IgG2a