missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ G-CSF Monoclonal Antibody (67604)
GREENER_CHOICE

Código de producto. 15513452
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
500 microgramos
Código de producto. Cantidad unitSize
15513452 500 μg 500 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15513452

Marca: Invitrogen™ MA523758

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

In ELISAs, does not cross-react with recombinant human (rh) G-CSF, rhCNTF, rmIL-6, rmLIF, or rmOSM. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet's syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno G-CSF
Aplicaciones ELISA, Neutralization, Western Blot, Western Blot
Clasificación Monoclonal
Clon 67604
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 5% trehalose and No Preservative
génica Csf3
N.º de referencia del gen P09920
Alias de gen C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4
Símbolos de los genes Csf3
Especie del huésped Rat
Inmunógeno E. coli-derived recombinant mouse G-CSF Val31-Ala208.
Método de purificación Protein A/G
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12985
Especies diana Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.