missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD6 Mouse anti-Human, Mouse, Rat, Pig, Rabbit, Chicken, Clone: 2H1B7, Proteintech
Tienda Proteintech ProductosDescripción
FXYD6 Monoclonal antibody apecifically detetects FXYD6 antigen in Human, Mouse, Rat, Pig, Rabbit, Chicken Samples. It is validated for Western Blot.
Especificaciones
Especificaciones
| Antígeno | FXYD6 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 2H1B7 |
| Concentración | 1000 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| N.º de referencia del gen | BC018652 |
| Símbolos de los genes | FXYD6 |
| Especie del huésped | Mouse |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?