missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUS/TLS Mouse anti-Human, Mouse, Clone: 3A10B5, Proteintech
Mouse Monoclonal Antibody
129.00€ - 359.47€
Especificaciones
| Antígeno | FUS/TLS |
|---|---|
| Clon | 3A10B5 |
| Concentración | 1 mg/mL |
| Aplicaciones | Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
Descripción
This antibody is a mouse monoclonal antibody raised against an internal region of human FUS. Initial reports from our customers suggest this new monoclonal FUS antibody (60160-1-Ig) is a useful tool in ALS and FTLD research. For more details, please see o
FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintece of healthy cells.Especificaciones
| FUS/TLS | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 75 kDa DNA pairing protein, FUS, FUS CHOP, FUS1, FUS-CHOP, hnRNP P2, Oncogene FUS, Oncogene TLS, POMp75, RNA binding protein FUS, TLS, TLS/CHOP | |
| FUS | |
| IgG1 | |
| Antigen Affinity Chromatography | |
| Antibody |
| 3A10B5 | |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| P35637, P56959 | |
| 233908, 2521 | |
| FUS/TLS Fusion Protein Ag2150 | |
| Primary | |
| -20°C | |
| FUS |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts