missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUS Recombinant Rabbit Monoclonal Antibody (BLR023E)

Descripción
Epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections.
This gene encodes a multifunctional protein component of the heterogeneous nuclear ribonucleoprotein (hnRNP) complex. The hnRNP complex is involved in pre-mRNA splicing and the export of fully processed mRNA to the cytoplasm. This protein belongs to the FET family of RNA-binding proteins which have been implicated in cellular processes that include regulation of gene expression, maintenance of genomic integrity and mRNA/microRNA processing. Alternative splicing results in multiple transcript variants. Defects in this gene result in amyotrophic lateral sclerosis type 6.
Especificaciones
Especificaciones
| Antígeno | FUS |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | BLR023E |
| Concentración | 30 μg/mL |
| Conjugado | Unconjugated |
| Formulación | BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 |
| génica | FUS |
| N.º de referencia del gen | P35637, P56959 |
| Alias de gen | 16) in malignant liposarcoma); 16) malignant liposarcoma; 75 kDa DNA-pairing protein; ALS6; D430004D17Rik; D930039C12Rik; ETM4; FUS; FUS RNA binding protein; Fus1; fused in sarcoma; fused in sarcoma RNA binding protein; fusion (involved in t(12; fusion gene in myxoid liposarcoma; fusion, derived from t(12; fusion, derived from t(12;16) malignant liposarcoma; fus-like protein; heterogeneous nuclear ribonucleoprotein P2; hnRNP P2; HNRNPP2; oncogene FUS; Oncogene TLS; pigpen protein; POMP75; Protein pigpen; RNA-binding protein FUS; TLS; translocated in liposarcoma; translocated in liposarcoma protein |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?