missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FPR3 Monoclonal Antibody (7D9B4)
GREENER_CHOICE

Código de producto. 17955451
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17955451 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17955451 Proveedor Invitrogen™ N.º de proveedor MA548461

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

FPR3 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

The N-formyl peptide receptor (FPR) family is comprised of three members, FPR, FPR3 (also designated FPRL1, lipoxin A4 receptor, FPRH1 or FPR2) and FPR like-2 (FPRL2), all of which are chemotactic G protein-coupled receptors that contain seven transmembrane domains. These receptors are found on the surface of phagocytic leukocytes, such as neutrophils and monocytes, and each family member contains specific residues, which are responsible for determining its ligand specificity. FPR3 is a promiscuous receptor that binds to several ligands, including lipoxin A4, N-formyl-methionyl-leucyl-phenylalanine (fMLP), serum amyloid A (SAA), prion peptide and the 42 amino acid form of beta amyloid. Upon activation, FPR3 induces migration and calcium mobilization in human monocytes and neutrophils and is involved in inflammatory and host defense responses. FPR3 may mediate inflammation in prion and Alzheimer's diseases, which makes it a potential target for therapeutic agents.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno FPR3
Aplicaciones ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Clasificación Monoclonal
Clon 7D9B4
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.05% sodium azide
génica FPR3
N.º de referencia del gen P25089
Alias de gen ALX; FML2_HUMAN; FMLP-related receptor I; FMLP-related receptor II; FMLP-R-I; FMLP-R-II; FMLPY; formyl peptide receptor 3; Formyl peptide receptor related sequence 1; Formyl peptide receptor-like 1; formyl peptide receptor-like 2; Formyl peptide receptor-related sequence 1; Fpr2; FPR3; FPRH1; FPRH2; Fprl1; FPRL2; Fpr-rs1; Fpr-s1; Lipoxin A4 receptor; LXA4 receptor; Lxa4r; LXA4-R; N-formyl peptide receptor 2; N-formyl peptide receptor 3; N-formylpeptide receptor-like 1; RGD1566044; RMLP-R-I
Símbolos de los genes FPR3
Especie del huésped Mouse
Inmunógeno Purified recombinant fragment of human FPR3 expressed in E. Coli.
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2359
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.