missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXRED1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA528507
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (85%), Pig (86%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that contains a FAD-dependent oxidoreductase domain. The encoded protein is localized to the mitochondria and may function as a chaperone protein required for the function of mitochondrial complex I. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants have been observed for this gene.
Especificaciones
| FOXRED1 | |
| Polyclonal | |
| Unconjugated | |
| FOXRED1 | |
| BC024806; FAD dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain-containing protein 1; Foxred1; FP634; H17; RGD1311785; TEG-23; testis expressed gene 23; Tex23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96CU9 | |
| FOXRED1 | |
| Recombinant protein encompassing a sequence within the center region of human FOXRED1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido