missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXA3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 11569090
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
11569090 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11569090

Marca: Invitrogen™ PA1813

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PA1-813 detects FOXA3 from human samples. PA1-813 has been successfully used in Western blot and ChIP applications. By Western blot, PA1-813 detects a ~37 kDa band representing FOXA3 from mouse liver. The PA1-813 immunogen is a synthetic peptide corresponding to residues S(4) V K M E A H D L A E W S Y Y P E A G E(23) of mouse FOXA3. This immunizing peptide (PEP-299) is available for use in neutralization and control experiments.

FOXA3 encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno FOXA3
Aplicaciones ChIP Assay, Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1 mg/mL BSA and 0.05% sodium azide
génica FOXA3
N.º de referencia del gen P32183, P35584, P55318
Alias de gen FKHH3; Fork head-related protein FKH H3; forkhead box A3; forkhead box protein A3; Foxa3; hepatocyte nuclear factor 3 gamma (winged helix transcription factor); hepatocyte nuclear factor 3, gamma; hepatocyte nuclear factor 3-gamma; HNF3G; HNF-3G; HNF-3-gamma; Tcf3g; Tcf-3g; Transcription factor 3G
Símbolos de los genes FOXA3
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to residues S(4) V K M E A H D L A E W S Y Y P E A G E(23) of mouse FOXA3.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 15377, 25100, 3171
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.