Learn More
FOXA2 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16881945 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16881945 150 μL 150 microlitros
16891945 20 μL 20 microlitros
2 options
Código de producto. 16881945
Marca: Proteintech 224741AP150UL
Rabbit Polyclonal Antibody
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
Especificaciones
Antígeno FOXA2
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica FOXA2
N.º de referencia del gen Q9Y261
Alias de gen forkhead box A2, Forkhead box protein A2, FOXA2, HNF 3 beta, HNF 3B, HNF3B, TCF 3B, TCF3B, Transcription factor 3B
Símbolos de los genes FOXA2
Especie del huésped Rabbit
Inmunógeno FOXA2 Fusion Protein Ag18221
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3170
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG