missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXA2 Monoclonal Antibody (7H4B7)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA515542
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-15542 targets FOXA2 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15542 immunogen is purified recombinant fragment of FOXA2 expressed in E. Coli. MA5-15542 detects FOXA2 which has a predicted molecular weight of approximately 48kDa.
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
Especificaciones
| FOXA2 | |
| Monoclonal | |
| Conc. not determined | |
| ascites with 0.03% sodium azide | |
| P35583, Q9Y261 | |
| FOXA2 | |
| Purified recombinant fragment of FOXA2 expressed in E. Coli. | |
| RUO | |
| 15376, 3170 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 7H4B7 | |
| Unconjugated | |
| FOXA2 | |
| FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido