Learn More
FOXA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16879944 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16879944 20 μL 20 microlitros
16869944 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16879944 Proveedor Proteintech N.º de proveedor 204111AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
Especificaciones
Antígeno FOXA1
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica FOXA1
N.º de referencia del gen P35582, P55317
Alias de gen forkhead box A1, Forkhead box protein A1, FOXA1, HNF 3 alpha, HNF 3A, HNF3A, TCF 3A, TCF3A, Transcription factor 3A
Símbolos de los genes FOXA1
Especie del huésped Rabbit
Inmunógeno FOXA1 Fusion Protein Ag14243
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 15375, 3169
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG