Learn More
- Home
- Products
- Tubos
- Tubos de plástico
- Cole-Parmer Food, Milk, and Dairy Tubing
Cole-Parmer Food, Milk, and Dairy Tubing
Código de producto. 30053077 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1-1/2 in
1-1/4 in
1-3/4 in
1/16 in
1/2 in
1/4 in
1/8 in
1 in
2 in
3/16 in
3/32 in
3/4 in
3/8 in
5/16 in
5/8 in
7/16 in
7/8 in
Diámetro exterior (imperial):
1-1/2 in
1-1/4 in
1-1/8 in
1-3/8 in
1/2 in
1/4 in
1/8 in
11/16 in
13/16 in
15/16 in
1 in
2-1/2 in
2-1/4 in
2 in
3/16 in
3/4 in
3/8 in
5/16 in
5/32 pulg
5/8 in
7/16 in
7/8 in
9/16 in
Tamaño de la unidad:
15 metros
| Código de producto. | Diámetro interior (imperial) | Diámetro exterior (imperial) | unitSize |
|---|---|---|---|
| 30053077 | 1/2 in | 3/4 in | 15 metros |
| 30053820 | 7/16 in | 9/16 in | 15 metros |
| 30053883 | 5/8 in | 7/8 in | 15 metros |
| 30054151 | 1 in | 1-1/4 in | 15 metros |
| 30054383 | 1/8 in | 3/16 in | 15 metros |
| 30053978 | 3/4 in | 1-1/4 in | 15 metros |
| 30053488 | 3/8 in | 5/8 in | 15 metros |
| 30054067 | 2 in | 2-1/2 in | 15 metros |
| 30054227 | 1/4 in | 3/8 in | 15 metros |
| 30054042 | 3/16 in | 1/4 in | 15 metros |
| 30053281 | 7/8 in | 1-1/8 in | 15 metros |
| 30053989 | 1/16 in | 1/8 in | 15 metros |
| 30054246 | 1/2 in | 11/16 in | 15 metros |
| 30054359 | 3/8 in | 9/16 in | 15 metros |
| 30053778 | 3/32 in | 5/32 pulg | 15 metros |
| 30053934 | 5/16 in | 9/16 in | 15 metros |
| 30053031 | 1-3/4 in | 2-1/4 in | 15 metros |
| 30053964 | 1/8 in | 1/4 in | 15 metros |
| 30054289 | 1 in | 1-1/2 in | 15 metros |
| 30054087 | 5/8 in | 15/16 in | 15 metros |
| 30052931 | 3/4 in | 1 in | 15 metros |
| 30053664 | 1-1/4 in | 1-1/2 in | 15 metros |
| 30053367 | 5/16 in | 7/16 in | 15 metros |
| 30054386 | 1/2 in | 5/8 in | 15 metros |
| 30054272 | 1-1/2 in | 2 in | 15 metros |
| 30053166 | 5/8 in | 13/16 in | 15 metros |
| 30054018 | 5/16 in | 1/2 in | 15 metros |
| 30054363 | 3/4 in | 1-1/8 in | 15 metros |
| 30053125 | 3/16 in | 7/16 in | 15 metros |
| 30053911 | 3/16 in | 5/16 in | 15 metros |
| 30053311 | 1 in | 1-3/8 in | 15 metros |
| 30053239 | 1/4 in | 1/2 in | 15 metros |
| 30053472 | 1/4 in | 7/16 in | 15 metros |
| 30054196 | 3/8 in | 1/2 in | 15 metros |
Este artículo no se puede devolver.
Vea la política de devoluciones
Código de producto. 30053077
Marca: Cole-Parmer 5012169
Inicie sesión o regístrese para comprobar su precio y disponibilidad.
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Crystal clear for visual inspection and flow monitoring. Non-toxic, odor-free, and taste-free. Smooth, non-porous and flexible tubing promotes a sanitary fluid path. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Especificaciones
| Diámetro interior (imperial) | 1/2 in |
| Diámetro exterior (imperial) | 3/4 in |
| Método de esterilización | No |
| Color | Clear |