missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FN3KRP Polyclonal Antibody

Descripción
Immunogen sequence: GSVLVMEHMDM RHLSSHAAKL GAQLADLHLD NKKLGEMRLK EAGTVGRGGG QEERPFVARF GFDVVTCCGY LPQVNDWQED WVVFYARQRI QPQMD Highest antigen sequence identity to the following orthologs - mouse 80%, rat 83%.
Fructosamine-3-kinase related protein (FN3KRP) is located on chromosome 17q25.3, approximately 8 kb upstream of the FN3K gene. It shares 65% amino acid identity with FN3K and possesses an identical genomic organization, implying functional similarities; FN3KRP is suggested to play a role in enzymatic deglycation. Unlike FN3K, FN3KRP is more ancient, present in diverse organisms such as cyanobacteria, echinoderms, plants, and cold-blooded vertebrates. Studies have discovered a significant association between rs1046896 in FN3KRP and longevity, where the longevity allele C is linked to increased FN3KRP expression in whole blood. FN3KRP is hypothesized to phosphorylate glucitolamines, products of non-enzymatic glycation, indicating its involvement in crucial metabolic processes. The regulation of both FN3K and FN3KRP is notable for TATA-less and CAAT-less promoters, featuring homologous CpG islands and Sp1 binding sites. Meanwhile, both genes are widely expressed across human tissues, but FN3K demonstrates significantly higher levels in tissues prone to glycation-related complications.
Especificaciones
Especificaciones
| Antígeno | FN3KRP |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | FN3KRP |
| N.º de referencia del gen | Q9HA64 |
| Alias de gen | FN3KL; FN3K-related protein; Fn3krp; FN3K-RP; fructosamine 3 kinase related protein; fructosamine 3-kinase-related protein; fructosamine-3-kinase-related protein; Ketosamine-3-kinase; ortholog of human fructosamine-3-kinase-related protein FN3KRP; Protein-psicosamine 3-kinase FN3KRP; RGD1304570; testis secretory sperm-binding protein Li 211a |
| Símbolos de los genes | FN3KRP |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?